Sodium thiosulfate 0.1 N in water

Supplier: BeanTown Chemical

137940-1L 137940-20L 137940-4L
BT137940-1LEA 44.28 USD
BT137940-1L BT137940-20L BT137940-4L
Sodium thiosulfate 0.1 N in water
Sodium thiosulphate

CAS: 7772-98-7; EC No: 231-867-5; MDL No: MFCD00003499 Liquid; Linear Formula: Na2S2O3; MW: 158.11 Density (g/mL): 1.01


  • Liquid

Formula: Na₂S₂O₃
Density: 1.01 g/mL
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR