1,3,5-Triisopropylbenzene ≥95%
Supplier: BeanTown Chemical
137925-1KG
137925-250G
137925-50G
BT137925-1KGEA
649
USD
BT137925-1KG
BT137925-250G
BT137925-50G
1,3,5-Triisopropylbenzene ≥95%
1,3,5-Triisopropylbenzene
CAS: 717-74-8; EC No: 211-941-3; MDL No: MFCD00008890
Liquid; Molecular Formula: C15H24; MW: 204.35
Boiling Point: 232-236°; Flash point: 87°C (189°F)
Density (g/mL): 0.845; Refractive Index: 1.488
- Refractive index: 1.488
- Liquid
Formula:
C₁₅H₂₄ Boiling Pt: 232…236 °C Density: 0.845 g/mL Flash Pt: 87 °C (189 °F) |
MDL Number:
MFCD00008890 CAS Number: 717-74-8 EINECS: 211-941-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...