Methyl (diethylphosphono)acetate ≥97%
Supplier: BeanTown Chemical
CAS: 1067-74-9; EC No: 213-938-2; MDL No: MFCD00009081
Liquid; Linear Formula: (CH3CH2O)2P(O)CH2COOCH3; Molecular Formula: C7H15O5P; MW: 210.17
Boiling Point: 127-131°/9 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.145; Refractive Index: 1.433
- Refractive index: 1.433
- Liquid
Formula:
C₇H₁₅O₅P Boiling Pt: 127…131 °C/9 mmHg Density: 1.145 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00009081 CAS Number: 1067-74-9 EINECS: 213-938-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...