Zinc meso-tetraphenylporphine

Supplier: BeanTown Chemical

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine Zinc(II), Zinc(II) Tetraphenylporphyrin, 5,10,15,20-Tetraphenyl-21H,23H-porphine zinc, Zinc TPP, Zinc(II) TPP, Tetraphenylporphine zinc, TPP Zn

137910-2G 137910-500MG
BT137910-2GEA 273 USD
BT137910-2G BT137910-500MG
Zinc meso-tetraphenylporphine
Zinc meso-tetraphenylporphine

CAS: 14074-80-7; MDL No: MFCD00012155 Crystalline; Molecular Formula: C44H28N4Zn ; MW: 678.10


  • Crystalline

Formula: C₄₄H₂₈N₄Zn
MW: 678.10 g/mol
MDL Number: MFCD00012155
CAS Number: 14074-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR