Diethyl (bromodifluoromethyl)phosphonate ≥97%
Supplier: BeanTown Chemical
137900-1G
137900-25G
137900-5G
BT137900-1GEA
26
USD
BT137900-1G
BT137900-25G
BT137900-5G
Diethyl (bromodifluoromethyl)phosphonate ≥97%
Diethyl (bromodifluoromethyl)phosphonate
CAS: 65094-22-6; MDL No: MFCD00069118
Liquid; Molecular Formula: C5H10BrF2O3P; MW: 267.01
Boiling Point: 40-41°/0.05 mmHg; Flash point:
- Refractive index: 1.417
- Liquid
Formula:
C₅H₁₀BrF₂O₃P Boiling Pt: 40…41 °C/0.05 mmHg Density: 1.503 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00069118 CAS Number: 65094-22-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...