Tin(IV) oxide ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Tin(iv)oxide, Stannic Oxide, Tin dioxide
137885-100G
137885-2KG
137885-500G
BT137885-100GEA
40
USD
BT137885-100G
BT137885-2KG
BT137885-500G
Tin(IV) oxide ≥99.9% (trace metals basis)
Tin(IV) oxide
CAS: 18282-10-5; EC No: 242-159-0; MDL No: MFCD00011244; RTECS: XQ4000000
- RTECS: XQ4000000
- Refractive index: 1.997
- <10 Micron Powder
Formula:
O₂Sn MW: 150.71 g/mol Boiling Pt: 1800…1900 °C Melting Pt: 1630 °C Density: 6.95 g/mL |
MDL Number:
MFCD00011244 CAS Number: 18282-10-5 EINECS: 242-159-0 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...