(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid ≥95%
Supplier: BeanTown Chemical
Synonyms:
Z-Dap-OH, Nα-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 3-Amino-N-(carbobenzoxy)-L-alanine, N(α)-Z-L-2,3-Diaminopropionic acid, (S)-Nalpha-Cbz-2,3-diaminopropionic Acid, Cbz-Dap-OH, Z-Dpr-OH, 3-Amino-N-Cbz-L-alanine
137850-1G
137850-5G
BT137850-1GEA
24
USD
BT137850-1G
BT137850-5G
(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid ≥95%
(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid
CAS: 35761-26-3; MDL No: MFCD00237345
Solid; Molecular Formula: C11H14N2O4; MW: 238.24
Melting Point: 240° (decomposes)
Optical Rotation: [α]20/D −39±1°, c = 1% in 1 M HCl
Hygroscopic
- Optical rotation: [α]20/D −39±1°, c = 1% in 1 M HCl
- Hygroscopic
- Solid
Formula:
C₁₁H₁₄N₂O₄ Melting Pt: 240 °C (dec.) |
MDL Number:
MFCD00237345 CAS Number: 35761-26-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...