Rhodium(II) acetate dimer ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

137830-1G 137830-250MG
BT137830-1GEA 605.55 USD
BT137830-1G BT137830-250MG
Rhodium(II) acetate dimer ≥99.99% (trace metals basis)
Rhodium (II) acetate dimer

CAS: 15956-28-2; EC No: 240-084-8; MDL No: MFCD00003538; RTECS: VI9361000 Powder; Linear Formula: Rh2(OOCCH3)4; MW: 441.99 Melting Point: 205°


  • RTECS: VI9361000
  • Powder

Formula: C₈H₁₂O₈Rh₂
MW: 441.987 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00003538
CAS Number: 15956-28-2
EINECS: 240-084-8

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR