Tetrabutylammonium bromide 98

Supplier: BeanTown Chemical

137810-100G 137810-2.5KG 137810-500G
BT137810-100GEA 57.76 USD
BT137810-100G BT137810-2.5KG BT137810-500G
Tetrabutylammonium bromide 98
Tetrabutylammonium bromide

CAS: 1643-19-2; EC No: 216-699-2; MDL No: MFCD00011633 Solid; Linear Formula: [CH3(CH2)3]4N(Br); Molecular Formula: C16H36BrN; MW: 322.38 Melting Point: 102-106° Density (g/mL): 1.15 Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 102…106 °C
Density: 1.15 g/mL
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR