Diethyl pyridine-3,5-dicarboxylate ≥96%

Supplier: BeanTown Chemical

Synonyms: Diethyl 3,5-Pyridinedicarboxylate, 3,5-Pyridinedicarboxylic acid diethyl ester

137795-1G 137795-5G
BT137795-1GEA 234 USD
BT137795-1G BT137795-5G
Diethyl pyridine-3,5-dicarboxylate ≥96%
Diethyl pyridine-3,5-dicarboxylate

CAS: 4591-56-4; EC No: 224-982-7; MDL No: MFCD00085755 Solid; Molecular Formula: C11H13NO4; MW: 223.23


  • Solid

Formula: C₁₁H₁₃NO₄
MW: 223.228 g/mol
MDL Number: MFCD00085755
CAS Number: 4591-56-4
EINECS: 224-982-7

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR