2-Amino-5-bromo-3-iodopyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-3-iodo-2-pyridylamine, 5-Bromo-3-iodopyridin-2-amine

137790-1G 137790-5G
BT137790-1GEA 159 USD
BT137790-1G BT137790-5G
2-Amino-5-bromo-3-iodopyridine ≥98%
2-Amino-5-bromo-3-iodopyridine

CAS: 381233-96-1; MDL No: MFCD06659000 Solid; Molecular Formula: C5H4BrIN2; MW: 298.91 Melting Point: 112-113° Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₅H₄BrIN₂
MW: 298.91 g/mol
Melting Pt: 112…113 °C
MDL Number: MFCD06659000
CAS Number: 381233-96-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR