6-Aminocaproic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid
137755-100G
137755-2.5KG
137755-500G
BT137755-100GEA
37.21
USD
BT137755-100G
BT137755-2.5KG
BT137755-500G
6-Aminocaproic acid ≥99%
6-Aminocaproic acid
CAS: 60-32-2; EC No: 200-469-3; MDL No: MFCD00008238; RTECS: MO6300000
Powder; Molecular Formula: C6H13NO2; MW: 131.17
Melting Point: 207-209° (decomposes)
- RTECS: MO6300000
- Powder
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Boiling Pt: 257 Melting Pt: 127 (dec.) Flash Pt: 207 |
MDL Number:
MFCD00008238 CAS Number: 60-32-2 EINECS: 200-469-3 Merck Index: 13,00431 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...