(-)-Di-p-toluoyl-L-tartaric acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid
137750-100G
137750-25G
137750-5G
BT137750-100GEA
455
USD
BT137750-100G
BT137750-25G
BT137750-5G
(-)-Di-p-toluoyl-L-tartaric acid ≥98%
(-)-Di-p-toluoyl-L-tartaric acid
CAS: 32634-66-5; EC No: 251-131-7; MDL No: MFCD00064440
Powder; Molecular Formula: C20H18O8; MW: 386.35
Melting Point: 169-171°
Optical Rotation: [α]20/D -138°, c = 1 in ethanol
- Optical rotation: [α]20/D -138°, c = 1 in ethanol
- Powder
Formula:
C₂₀H₁₈O₈ Melting Pt: 169…171 °C |
MDL Number:
MFCD00064440 CAS Number: 32634-66-5 EINECS: 251-131-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...