1,2-Bis(tosyloxy)ethane ≥97%

Supplier: BeanTown Chemical

Synonyms: Ethane-1,2-diyl bis(4-methylbenzenesulphonate), Ethylene Glycol Ditosylate, 1,2-Bis(p-toluenesulphonyloxy)ethane, Ethylene Glycol Bis(p-toluenesulphonate), Ethylene glycol bis-p-toluenesulphonate

137705-10G 137705-50G
BT137705-10GEA 48 USD
BT137705-10G BT137705-50G
1,2-Bis(tosyloxy)ethane ≥97%
1,2-Bis(tosyloxy)ethane

CAS: 6315-52-2; MDL No: MFCD00008549 Solid; Linear Formula: [CH3C6H4SO3CH2]2; Molecular Formula: C16H18O6S2; MW: 370.45 Melting Point: 122-127°


  • Solid

Formula: C₁₆H₁₈O₆S₂
Melting Pt: 122…127 °C
MDL Number: MFCD00008549
CAS Number: 6315-52-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR