(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein ≥97%

Supplier: BeanTown Chemical

Synonyms: (E)-3-[3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl]-propenal, (E)-3-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)acrylaldehyde, (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]propenal

137685-25G 137685-5G
BT137685-25GEA 464 USD
BT137685-25G BT137685-5G
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein ≥97%
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein

CAS: 93957-50-7; EC No: 425-370-4; MDL No: MFCD03840863 Solid; Molecular Formula: C20H18FNO; MW: 307.36 Density (g/mL): 1.10


  • Solid

Formula: C₂₀H₁₈FNO
MW: 307.37 g/mol
Density: 1.10 g/mL
MDL Number: MFCD03840863
CAS Number: 93957-50-7
EINECS: 425-370-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR