Methyl-3-aminocrotonate ≥97%

Supplier: BeanTown Chemical

Synonyms: Methyl 3-Amino-2-butenoate, 3-Aminocrotonic acid Methyl Ester, Methyl 3-aminobut-2-enoate, 3-Amino-2-butenoic acid Methyl Ester, Methyl 3-Aminocrotonate

137675-100G 137675-500G
BT137675-100GEA 35 USD
BT137675-100G BT137675-500G
Methyl-3-aminocrotonate ≥97%
Methyl 3-aminocrotonate

CAS: 14205-39-1; EC No: 238-056-5; MDL No: MFCD00008072; RTECS: EM9092500 Crystalline; Linear Formula: CH3C(NH2)=CHCOOCH3; Molecular Formula: C5H9NO2 ; MW: 115.13 Melting Point: 81-83°; Boiling Point: 112°/42 mmHg; Flash point: 91°C (196°F) Hygroscopic


  • RTECS: EM9092500
  • Hygroscopic
  • Crystalline

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 112 °C/42 mmHg
Melting Pt: 81…83 °C
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00008072
CAS Number: 14205-39-1
EINECS: 238-056-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR