Bis(phenylacetyl)disulfide ≥98%
Supplier: BeanTown Chemical
137670-25G
137670-5G
BT137670-25GEA
102
USD
BT137670-25G
BT137670-5G
Bis(phenylacetyl)disulfide ≥98%
Bis(phenylacetyl)disulfide
CAS: 15088-78-5; MDL No: MFCD00513572
Solid; Molecular Formula: C16H14O2S2; MW: 302.42
Melting Point: 58-60°
- Solid
Formula:
C₁₆H₁₄O₂S₂ Melting Pt: 58…60 °C |
MDL Number:
MFCD00513572 CAS Number: 15088-78-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...