L(+)-Arabinose ≥99%
Supplier: BeanTown Chemical
Synonyms:
Pectinose, Pectin Sugar
137665-250G
137665-50G
BT137665-250GEA
310.49
USD
BT137665-250G
BT137665-50G
L(+)-Arabinose ≥99%
L(+)-Arabinose
EC No: 201-767-6
Solid
Optical Rotation: [α]20/D +103°, c = 1 in water
- Optical rotation: [α]20/D +103°, c = 1 in water
- Solid
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Melting Pt: 160-163 °C |
MDL Number:
MFCD00067709 CAS Number: 5328-37-0 EINECS: 201-767-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...