Monosodium 5-sulfoisophthalate ≥95%

Supplier: BeanTown Chemical

Synonyms: Sodium 3,5-dicarboxybenzenesulphonate, 5-Sulphoisophthalic acid monosodium salt, Sodium 5-sulphoisophthalate

137660-100G 137660-500G
BT137660-100GEA 29 USD
BT137660-100G BT137660-500G
Monosodium 5-sulfoisophthalate ≥95%
Monosodium 5-sulfoisophthalate

CAS: 6362-79-4; EC No: 228-845-2; MDL No: MFCD00007495; RTECS: DB6030000 Powder; Molecular Formula: C8H5NaO7S ; MW: 268.18


  • RTECS: DB6030000
  • Powder

Formula: C₈H₆O₇S
MW: 268.17 g/mol
MDL Number: MFCD00007495
CAS Number: 6362-79-4
EINECS: 228-845-2

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR