Dichlorobis(ethylenediamine)platinum(II)
Supplier: BeanTown Chemical
137645-1G
137645-250MG
BT137645-1GEA
234
USD
BT137645-1G
BT137645-250MG
Dichlorobis(ethylenediamine)platinum(II)
Dichlorobis(ethylenediamine)platinum(II)
CAS: 21430-85-3; MDL No: MFCD00009881
Crystalline; Molecular Formula: C4H16Cl2N4Pt; MW: 386.18
Melting Point: 300° (decomposes)
- Crystalline
Formula:
C₄H₁₆Cl₂N₄Pt Melting Pt: 300 °C (dec.) |
MDL Number:
MFCD00009881 CAS Number: 21430-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...