Phenylglyoxal monohydrate ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone
137630-100G
137630-25G
137630-5G
BT137630-100GEA
824
USD
BT137630-100G
BT137630-25G
BT137630-5G
Phenylglyoxal monohydrate ≥97%
Phenylglyoxal monohydrate
CAS: 1075-06-5; EC No: 214-036-1; MDL No: MFCD00149499; RTECS: KM5775180
Solid; Linear Formula: C6H5COCHO·H2O; Molecular Formula: C8H6O2·H2O; MW: 152.15
Melting Point: 71-77°; Boiling Point: 142°/125 mmHg
- RTECS: KM5775180
- Solid
Formula:
C₈H₆O₂·H₂O Boiling Pt: 142 °C/125 mmHg Melting Pt: 71…77 °C |
MDL Number:
MFCD00149499 CAS Number: 1075-06-5 EINECS: 214-036-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...