1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Supplier: BeanTown Chemical
137610-2G
137610-500MG
BT137610-2GEA
403
USD
BT137610-2G
BT137610-500MG
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
CAS: 38465-86-0; MDL No: MFCD00064800
Crystalline; Molecular Formula: C34H38F6IrP3; MW: 845.79
Melting Point: 224° (decomposes)
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₃₄H₃₈F₆IrP₃ Melting Pt: 224 °C (dec.) |
MDL Number:
MFCD00064800 CAS Number: 38465-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...