cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate
Supplier: BeanTown Chemical
Synonyms:
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) dihydrate
137595-1G
137595-5G
BT137595-1GEA
111
USD
BT137595-1G
BT137595-5G
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate
CAS: 15746-57-3; MDL No: MFCD00012040
Crystalline; Molecular Formula: C20H16Cl2N4Ru·2H2O; MW: 520.38
- Crystalline
Formula:
C₂₀H₂₀Cl₂N₄O₂Ru MW: 520.376 g/mol |
MDL Number:
MFCD00015504 CAS Number: 15746-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...