Octafluoronaphthalene ≥96%
Supplier: BeanTown Chemical
Synonyms:
Perfluoronaphthalene
137570-1G
137570-25G
137570-5G
BT137570-1GEA
45
USD
BT137570-1G
BT137570-25G
BT137570-5G
Octafluoronaphthalene ≥96%
Octafluoronaphthalene
CAS: 313-72-4; EC No: 206-239-9; MDL No: MFCD00014307
Solid; Molecular Formula: C10F8 ; MW: 272.10
Melting Point: 87-88°; Boiling Point: 209°
- Solid
Formula:
C₁₀F₈ MW: 272.1 g/mol Boiling Pt: 209 °C Melting Pt: 87…88 °C |
MDL Number:
MFCD00014307 CAS Number: 313-72-4 EINECS: 206-239-9 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...