1,3-Dimethylbarbituric acid, (max. 6% H₂O) ≥99% (dry weight)

Supplier: BeanTown Chemical

137565-1KG 137565-250G 137565-50G
BT137565-1KGEA 410.82 USD
BT137565-1KG BT137565-250G BT137565-50G
1,3-Dimethylbarbituric acid, (max. 6% H₂O) ≥99% (dry weight)
1,3-Dimethylbarbituric acid

CAS: 769-42-6; EC No: 212-211-7; MDL No: MFCD00006675 Crystalline; Molecular Formula: C6H8N2O3; MW: 156.14 Melting Point: 121-123°


  • Crystalline

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00006675
CAS Number: 769-42-6
EINECS: 212-211-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR