4-Methylumbelliferone ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-MU
137560-1KG
137560-250G
137560-50G
BT137560-1KGEA
304
USD
BT137560-1KG
BT137560-250G
BT137560-50G
4-Methylumbelliferone ≥97%
4-Methylumbelliferone
CAS: 90-33-5; EC No: 201-986-7; MDL No: MFCD00006866; RTECS: GN7000000
Solid; Molecular Formula: C10H8O3 ; MW: 176.17
- RTECS: GN7000000
- Solid
Formula:
C₁₀H₈O₃ MW: 176.17 g/mol |
MDL Number:
MFCD00006866 CAS Number: 90-33-5 EINECS: 201-986-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...