Praseodymium(III,IV) oxide ≥99.99%

Supplier: BeanTown Chemical

137555-100G 137555-25G
BT137555-100GEA 879 USD
BT137555-100G BT137555-25G
Praseodymium(III,IV) oxide ≥99.99%
Praseodymium(III,IV) oxide

CAS: 12037-29-5; EC No: 234-857-9; MDL No: MFCD00011178 Powder; Linear Formula: Pr6O11; MW: 1021.43 Melting Point: 2500° Density (g/mL): 6.5


  • Powder

Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2500 °C
Density: 6.5 g/mL
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR