2-Aminobenzenesulfonic acid ≥97%
Supplier: BeanTown Chemical
137530-100G
137530-500G
BT137530-100GEA
49
USD
BT137530-100G
BT137530-500G
2-Aminobenzenesulfonic acid ≥97%
2-Aminobenzenesulphonic acid
CAS: 88-21-1; EC No: 201-810-9; MDL No: MFCD00007705; RTECS: DB4727700
Powder; Linear Formula: C6H4(NH2)(SO3H); MW: 173.19
Melting Point: 300°
- RTECS: DB4727700
- Powder
Formula:
C₆H₇NO₃S Melting Pt: 300 °C |
MDL Number:
MFCD00007705 CAS Number: 88-21-1 EINECS: 201-810-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...