2'-Bromo-4'-fluoroacetophenone ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-(2-Bromo-4-fluorophenyl)ethanone

137510-1G 137510-5G
BT137510-1GEA 75 USD
BT137510-1G BT137510-5G
2'-Bromo-4'-fluoroacetophenone ≥98%
2'-Bromo-4'-fluoroacetophenone

CAS: 1006-39-9; MDL No: MFCD00077464 Liquid; Molecular Formula: C8H6BrFO; MW: 217.04


  • Liquid

Formula: C₈H₆BrFO
MW: 217.04 g/mol
MDL Number: MFCD00077464
CAS Number: 1006-39-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR