(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer

Supplier: BeanTown Chemical
Warning

Synonyms: Dichloro(pentamethylcyclopentadienyl)iridium(III) Dimer, Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, [Ir(Cp-)Cl2]2, (Pentamethylcyclopentadienyl)iridium (III) chloride dimer

137470-2G 137470-500MG
BT137470-2GEA 522 USD
BT137470-2G BT137470-500MG
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer

CAS: 12354-84-6; MDL No: MFCD00075435 Solid; Molecular Formula: C20H30Cl4Ir2; MW: 796.67


  • Solid

Formula: C₂₀H₃₀Cl₄Ir₂
MW: 796.69 g/mol
MDL Number: MFCD00075435
CAS Number: 12354-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR