4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥97%

Supplier: BeanTown Chemical

137395-1G 137395-5G
BT137395-1GEA 103 USD
BT137395-1G BT137395-5G
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥97%
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate

CAS: 123843-69-6; MDL No: MFCD11053375 Solid; Molecular Formula: C19H17F2NO2; MW: 329.35 Melting Point: 45°


  • Solid

Formula: C₁₉H₁₇F₂NO₂
Melting Pt: 45 °C
MDL Number: MFCD11053375
CAS Number: 123843-69-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR