N-Carbobenzoxy-6-aminohexanoic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: N-Cbz-epsilon-aminocaproic Acid, N-Cbz-6-aminohexanoic Acid, N-Benzyloxycarbonyl-6-aminohexanoic acid, N-Carbobenzoxy-epsilon-aminocaproic Acid, Z-ε-Acp-OH

137360-25G 137360-5G
BT137360-25GEA 138 USD
BT137360-25G BT137360-5G
N-Carbobenzoxy-6-aminohexanoic acid ≥99%
N-Carbobenzoxy-6-aminohexanoic acid

CAS: 1947-00-8; EC No: 217-748-0; MDL No: MFCD00004423 Powder; Linear Formula: C6H5CH2O2CNH(CH2)5COOH; Molecular Formula: C14H19NO4; MW: 265.31 Melting Point: 54-57°; Flash point: 110°C (230°F)


  • Powder

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Melting Pt: 54…57 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004423
CAS Number: 1947-00-8
EINECS: 217-748-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR