2-Chloro-3-iodopyridine ≥97%

Supplier: BeanTown Chemical

137325-1G 137325-5G
BT137325-1GEA 80 USD
BT137325-1G BT137325-5G
2-Chloro-3-iodopyridine ≥97%
2-Chloro-3-iodopyridine

CAS: 78607-36-0; MDL No: MFCD00661298 Solid; Molecular Formula: C5H3ClIN ; MW: 239.44 Melting Point: 90-95° Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₅H₃ClIN
MW: 239.44 g/mol
Melting Pt: 90…95 °C
MDL Number: MFCD00661298
CAS Number: 78607-36-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR