Dopamine hydrochloride ≥99%
Supplier: BeanTown Chemical
137320-100G
137320-25G
137320-5G
BT137320-100GEA
393.6
USD
BT137320-100G
BT137320-25G
BT137320-5G
Dopamine hydrochloride ≥99%
Dopamine hydrochloride
CAS: 62-31-7; EC No: 200-527-8; MDL No: MFCD00012898; RTECS: UX1092000
Powder; Molecular Formula: C8H11NO2·HCl; MW: 189.64
Melting Point: 248-250°
Hygroscopic , Light Sensitive
- RTECS: UX1092000
- Hygroscopic, Light Sensitive
- Powder
Formula:
C₈H₁₂ClNO₂ Melting Pt: 248…250 °C |
MDL Number:
MFCD00012898 CAS Number: 62-31-7 EINECS: 200-527-8 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...