Calix[4]arene ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,3,5,7(1,3)-Tetrabenzenacyclooctaphane-12,32,52,72-tetraol
137315-1G
137315-5G
BT137315-1GEA
79
USD
BT137315-1G
BT137315-5G
Calix[4]arene ≥98%
Calix[4]arene
CAS: 74568-07-3; MDL No: MFCD00143912
Powder; Molecular Formula: C28H24O4; MW: 424.49
- Powder
Formula:
C₂₈H₂₄O₄ MW: 424.5 g/mol |
MDL Number:
MFCD00233673 CAS Number: 74568-07-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...