2-Decanone ≥97%

Supplier: BeanTown Chemical

Synonyms: Decan-2-one

137290-100G 137290-25G 137290-5G
BT137290-100GEA 193 USD
BT137290-100G BT137290-25G BT137290-5G
2-Decanone ≥97%
2-Decanone

CAS: 693-54-9; EC No: 211-752-6; MDL No: MFCD00009571; RTECS: HE0725000 Liquid; Linear Formula: CH3(CH2)7COCH3; Molecular Formula: C10H20O; MW: 156.27 Melting Point: 3°; Boiling Point: 211°; Flash point: 86°C (187°F) Density (g/mL): 0.825; Refractive Index: 1.425


  • RTECS: HE0725000
  • Refractive index: 1.425
  • Liquid

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 211 °C
Melting Pt: 3 °C
Density: 0.825 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00009571
CAS Number: 693-54-9
EINECS: 211-752-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR