2-(Chloromethyl)benzimidazole ≥96%
Supplier: BeanTown Chemical
137250-25G
137250-5G
BT137250-25GEA
141
USD
BT137250-25G
BT137250-5G
2-(Chloromethyl)benzimidazole ≥96%
2-(Chloromethyl)benzimidazole
CAS: 4857-04-9; EC No: 225-452-8; MDL No: MFCD00005602
Powder; Molecular Formula: C8H7ClN2; MW: 166.61
Melting Point: 146-148° (decomposes)
- Powder
Formula:
C₈H₇ClN₂ Melting Pt: 146…148 °C (dec.) |
MDL Number:
MFCD00005602 CAS Number: 4857-04-9 EINECS: 225-452-8 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...