Di-tert-butyl phosphonate ≥95%
Supplier: BeanTown Chemical
Synonyms:
Di-tert-butyl phosphite
137225-1G
137225-25G
137225-5G
BT137225-1GEA
30
USD
BT137225-1G
BT137225-25G
BT137225-5G
Di-tert-butyl phosphonate ≥95%
Di-tert-butyl phosphonate
CAS: 13086-84-5; EC No: 235-996-8; MDL No: MFCD00014999
Liquid; Molecular Formula: C8H19O3P; MW: 194.21
Density (g/mL): 0.96; Refractive Index: 1.419
Air Sensitive, Moisture Sensitive
- Refractive index: 1.419
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₈H₁₉O₃P MW: 194.21 Density: 0.96 g/mL |
MDL Number:
MFCD00995019 CAS Number: 13086-84-5 EINECS: 235-996-8 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...