Butyl stearate, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Stearic acid butyl ester, Butyl octadecanoate

137215-2KG 137215-500G
BT137215-2KGEA 103 USD
BT137215-2KG BT137215-500G
Butyl stearate, Technical Grade
Butyl stearate

CAS: 123-95-5; EC No: 204-666-5; MDL No: MFCD00026669; RTECS: WI2900000 Liquid; Linear Formula: CH3(CH2)16COO(CH2)3CH3 ; Molecular Formula: C22H44O2; MW: 340.58 Melting Point: 17-22°; Flash point: 160°C (320°F) Density (g/mL): 0.861; Refractive Index: 1.443


  • RTECS: WI2900000
  • Refractive index: 1.443
  • Liquid

Formula: C₂₂H₄₄O₂
MW: 340.59 g/mol
Boiling Pt: 220
Melting Pt: 21
Density: 0.861 g/mL
Flash Pt: 160 °C (320 °F)
MDL Number: MFCD00026669
CAS Number: 123-95-5
EINECS: 204-666-5
Merck Index: 13,01589

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR