Bismuth(III) oxide ≥99.95%

Supplier: BeanTown Chemical

Synonyms: Bismuth Trioxide, Dibismuth trioxide

137195-1KG 137195-250G 137195-50G
BT137195-1KGEA 359 USD
BT137195-1KG BT137195-250G BT137195-50G
Bismuth(III) oxide ≥99.95%
Bismuth(III) oxide

CAS: 1304-76-3; EC No: 215-134-7; MDL No: MFCD00003462; RTECS: EB2984460 Powder, typically 10 micron ; Molecular Formula: Bi2O3; MW: 465.96


  • RTECS: EB2984460
  • Powder, typically <10 micron

Formula: Bi₂O₃
MW: 465.96 g/mol
Boiling Pt: 1890
Melting Pt: 825
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR