4-Methoxyphenylacetone ≥98%

Supplier: BeanTown Chemical

137190-100G 137190-25G 137190-500G
BT137190-100GEA 209 USD
BT137190-100G BT137190-25G BT137190-500G
4-Methoxyphenylacetone ≥98%
4-Methoxyphenylacetone

CAS: 122-84-9; EC No: 204-578-7; MDL No: MFCD00008773; RTECS: UC2990000 Liquid; Molecular Formula: C10H12O2 ; MW: 164.20 Boiling Point: 145°/25 mmHg; Flash point: 102°C (216°F) Density (g/mL): 1.067; Refractive Index: 1.525


  • RTECS: UC2990000
  • Refractive index: 1.525
  • Liquid

Formula: C₁₀H₁₂O₂
Boiling Pt: 145 °C/25 mmHg
Density: 1.067 g/mL
Flash Pt: 102 °C (216 °F)
MDL Number: MFCD00008773
CAS Number: 122-84-9
EINECS: 204-578-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR