Duloxetine hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
(S)-(+)-N-Methyl-3-(1-naphthyloxy)-3-(2-thienyl)propylamine Hydrochloride, (S)-Duloxetine hydrochloride
137145-10MG
137145-50MG
BT137145-10MGEA
52
USD
BT137145-10MG
BT137145-50MG
Duloxetine hydrochloride ≥98%
Duloxetine hydrochloride
CAS: 136434-34-9; MDL No: MFCD06407958; RTECS: XN0258000
Solid; Molecular Formula: C18H19NOS·HCl; MW: 333.88
Melting Point: 118-122°
Air Sensitive
- RTECS: XN0258000
- Air Sensitive
- Solid
Formula:
C₁₈H₁₉NOS Melting Pt: 118…122 °C |
MDL Number:
MFCD06407958 CAS Number: 136434-34-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...