Aluminium oxide, α-phase ≥99.98% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: ɑ-Tonerde, α-Alumina

137075-100G 137075-25G 137075-500G
BT137075-100GEA 52.7 USD
BT137075-100G BT137075-25G BT137075-500G
Aluminium oxide, α-phase ≥99.98% (trace metals basis)
Aluminium oxide, α-phase

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 0.80-1.0 Micron APS Powder, S.A. 3.0-4.0m2/g ; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • 0.80-1.0 Micron APS Powder, S.A. 3.0-4.0m²/g

Formula: Al₂O₃
MW: 101.96 g/mol
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Assay 99.98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR