1,3-Di-tert-butylbenzene ≥99%

Supplier: BeanTown Chemical

137070-1G 137070-25G 137070-5G
BT137070-1GEA 68 USD
BT137070-1G BT137070-25G BT137070-5G
1,3-Di-tert-butylbenzene ≥99%
1,3-Di-tert-butylbenzene

CAS: 1014-60-4; MDL No: MFCD00008830 Liquid; Molecular Formula: C14H22; MW: 190.32 Melting Point: 10-11°; Boiling Point: 106-107°/18 mmHg; Flash point: 84°C (183°F) Density (g/mL): 0.859; Refractive Index: 1.488


  • Refractive index: 1.488
  • Liquid

Formula: C₁₄H₂₂
MW: 190.33
Boiling Pt: 106…107 °C/18 mmHg
Melting Pt: 10…11 °C
Density: 0.859 g/mL
Flash Pt: 84 °C (183 °F)
MDL Number: MFCD00008830
CAS Number: 1014-60-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR