trans-4-(4-Propylcyclohexyl)benzonitrile ≥99%
Supplier: BeanTown Chemical
Synonyms:
4-(trans-4-propylcyclohexyl)benzonitrile
137065-1G
137065-5G
BT137065-1GEA
83
USD
BT137065-1G
BT137065-5G
trans-4-(4-Propylcyclohexyl)benzonitrile ≥99%
trans-4-(4-Propylcyclohexyl)benzonitrile
CAS: 61203-99-4; EC No: 262-654-5; MDL No: MFCD04038799
Solid; Molecular Formula: C16H21N; MW: 227.35
Melting Point: 45°
- Solid
Formula:
C₁₆H₂₁N MW: 227.3446 g/mol Melting Pt: 45 °C |
MDL Number:
MFCD04038799 CAS Number: 61203-99-4 EINECS: 262-654-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...