1,2-Cyclohexanedione ≥98%
Supplier: BeanTown Chemical
137055-25G
137055-5G
BT137055-25GEA
228
USD
BT137055-25G
BT137055-5G
1,2-Cyclohexanedione ≥98%
1,2-Cyclohexanedione
CAS: 765-87-7; EC No: 212-155-3; MDL No: MFCD00001648; RTECS: GV0340000
Solid; Molecular Formula: C6H8O2; MW: 112.13
Melting Point: 34-38°; Boiling Point: 193-195°; Flash point: 84°C (183°F)
Density (g/mL): 1.12
- RTECS: GV0340000
- Solid
Formula:
C₆H₈O₂ Boiling Pt: 193…195 °C Melting Pt: 34…38 °C Density: 1.12 g/mL |
MDL Number:
MFCD00001648 CAS Number: 765-87-7 EINECS: 212-155-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...