2-Naphthyl boronic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid
137050-1G
137050-25G
137050-5G
BT137050-1GEA
26
USD
BT137050-1G
BT137050-25G
BT137050-5G
2-Naphthyl boronic acid ≥97%
2-Naphthyl boronic acid
CAS: 32316-92-0; MDL No: MFCD00236051
Powder; Molecular Formula: C10H9BO2 ; MW: 171.99
Melting Point: 269-275°
- Powder
Formula:
C₁₀H₉BO₂ Melting Pt: 269…275 °C |
MDL Number:
MFCD00236051 CAS Number: 32316-92-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...