(2-Bromoethoxy)benzene ≥98%

Supplier: BeanTown Chemical

Synonyms: β-Bromophenetole, 2-Phenoxyethyl bromide, 2-Bromoethyl phenyl ether, ω-Bromophenetole

137045-100G 137045-25G 137045-500G
BT137045-100GEA 122 USD
BT137045-100G BT137045-25G BT137045-500G
(2-Bromoethoxy)benzene ≥98%
(2-Bromoethoxy)benzene

CAS: 589-10-6; EC No: 209-634-4; MDL No: MFCD00000234 Crystalline; Molecular Formula: C8H9BrO ; MW: 201.06 Melting Point: 31-34°; Boiling Point: 144°/40 mmHg; Flash point: 65°C (149°F)


  • Crystalline

Formula: BrCH₂CH₂OC₆H₅
MW: 201.06 g/mol
Boiling Pt: 144
Melting Pt: 31…34 °C
Flash Pt: 65 °C (149 °F)
MDL Number: MFCD00000234
CAS Number: 589-10-6
EINECS: 209-634-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR