Ethyl (R)-(-)-3-hydroxybutyrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl (R)-3-hydroxybutyrate, (R)-(-)-3-Hydroxybutyric Acid Ethyl Ester

137030-1G 137030-5G
BT137030-1GEA 47 USD
BT137030-1G BT137030-5G
Ethyl (R)-(-)-3-hydroxybutyrate ≥98%
Ethyl (R)-(-)-3-hydroxybutyrate

CAS: 24915-95-5; MDL No: MFCD00075386 Liquid; Molecular Formula: C6H12O3 ; MW: 132.16 Boiling Point: 75-76°/12 mmHg; Flash point: 64°C (147°F) Density (g/mL): 1.017; Refractive Index: 1.420; Optical Rotation: [α]20/D −46°, c = 1 in chloroform


  • Optical rotation: [α]20/D −46°, c = 1 in chloroform
  • Refractive index: 1.420
  • Liquid

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 75…76 °C/12 mmHg
Density: 1.017 g/mL
Flash Pt: 64 °C (147 °F)
MDL Number: MFCD00075386
CAS Number: 24915-95-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR