Platinum iridium alloy (80:20 w%), wire, hard type, Diameter 0.5 mm

Supplier: BeanTown Chemical

136995-10CM 136995-50CM
BT136995-10CMEA 125 USD
BT136995-10CM BT136995-50CM
Platinum iridium alloy (80:20 w%), wire, hard type, Diameter 0.5 mm
Platinum iridium alloy (80:20 w%)

MDL No: MFCD02091683 Solid; Formula: Pt:Ir; 80:20 wt%


  • Solid

MDL Number: MFCD02091683
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR